Upadacitinib 99%

Supplier: Ambeed
Warning

Synonyms: ABT-494, (3S,4R)-3-ethyl-4-(3H-imidazo[1,2-a]pyrrolo[2,3-e]pyrazin-8-yl)-N-(2,2,2-trifluoroethyl)pyrrolidine-1-carboxamide

A165008-10MG A165008-1MG A165008-25MG A165008-5MG A165008-100MG A165008-250MG A165008-5G A165008-50MG
77247-756EA 56.65 USD
77247-756 77247-758 77247-760 77247-762 77615-354 77615-356 77746-114 77746-113
Upadacitinib 99%
Upadacitinib

Precaución: for Research Use Only

Formula: C₁₇H₁₉F₃N₆O
MW: 380.37 g/mol
MDL Number: MFCD30502663
CAS Number: 1310726-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR