3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate 97%

Supplier: Ambeed
Danger

A164997-10G A164997-25G A164997-5G
77273-500EA 32 USD
77273-500 77273-502 77273-504
3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate 97%
3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate

Precaución: for Research Use Only

Formula: C₈H₉N₃S·H₂O·HCl
MDL Number: MFCD00149370
CAS Number: 38894-11-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR