5-Bromo-2,4-difluoroaniline 98%

Supplier: Ambeed
Warning

A164904-100G A164904-10G A164904-25G A164904-5G
77286-574EA 119.48 USD
77286-574 77286-576 77286-578 77286-580
5-Bromo-2,4-difluoroaniline 98%
5-Bromo-2,4-difluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄BrF₂N
MW: 208.0000 g/mol
MDL Number: MFCD03426368
CAS Number: 452-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR