9-Hexyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole 97%

Supplier: Ambeed
Warning

A164699-1G A164699-250MG A164699-5G
77236-030EA 33.99 USD
77236-030 77236-032 77236-034
9-Hexyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole 97%
9-Hexyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole

Precaución: for Research Use Only

Formula: C₃₀H₄₃B₂NO₄
MW: 503.30 g/mol
MDL Number: MFCD30187269
CAS Number: 871696-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR