2-Benzyloxyphenol 98%
Supplier: Ambeed
|
Warning
|
A164557-100G
A164557-10G
A164557-1G
A164557-5G
77236-022EA
145
USD
77236-022
77236-024
77236-026
77236-028
2-Benzyloxyphenol 98%
2-Benzyloxyphenol
Precaución: for Research Use Only
Formula:
C₁₃H₁₂O₂ |
MDL Number:
MFCD00002186 CAS Number: 6272-38-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...