2-(Difluoromethoxy)benzonitrile 95%

Supplier: Ambeed
Warning

A164556-1G A164556-250MG A164556-5G
77236-016EA 14 USD
77236-016 77236-018 77236-020
2-(Difluoromethoxy)benzonitrile 95%
2-(Difluoromethoxy)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₅F₂NO
MDL Number: MFCD00221469
CAS Number: 56935-78-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR