5-Benzyloxy-2-bromo-4-methoxybenzaldehyde 97%

Supplier: Ambeed
Warning

A164425-1G A164425-250MG
77247-746EA 70.04 USD
77247-746 77247-748
5-Benzyloxy-2-bromo-4-methoxybenzaldehyde 97%
5-Benzyloxy-2-bromo-4-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₃BrO₃
MW: 321.17 g/mol
MDL Number: MFCD00666893
CAS Number: 6451-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR