(2Z,3E)-3-(Hydroxyimino)-[2,3'-biindolinylidene]-2'-one 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Indirubin-3′-oxime
A164297-100MG
A164297-250MG
A164297-25MG
A164297-50MG
77259-734EA
125.66
USD
77259-734
77259-736
77259-738
77259-740
(2Z,3E)-3-(Hydroxyimino)-[2,3'-biindolinylidene]-2'-one 98%
(2Z,3E)-3-(Hydroxyimino)-[2,3'-biindolinylidene]-2'-one
Precaución: for Research Use Only
Formula:
C₁₆H₁₁N₃O₂ MW: 277.27 g/mol |
MDL Number:
MFCD02683594 CAS Number: 667463-82-3 UN: 1325 ADR: 4.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...