2,3,4-Trimethoxyphenol 97%

Supplier: Ambeed
Warning

A164247-100MG A164247-1G A164247-250MG
77247-740EA 23 USD
77247-740 77247-742 77247-744
2,3,4-Trimethoxyphenol 97%
2,3,4-Trimethoxyphenol

Precaución: for Research Use Only

Formula: C₉H₁₂O₄
MW: 184.19 g/mol
MDL Number: MFCD00068804
CAS Number: 19676-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR