3-Chloropropyl benzoate 97%

Supplier: Ambeed
Warning

A164173-100MG A164173-250MG
77247-736EA 53.56 USD
77247-736 77247-738
3-Chloropropyl benzoate 97%
3-Chloropropyl benzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₂
MW: 198.64 g/mol
MDL Number: MFCD20621205
CAS Number: 942-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR