(3-Chloropropyl)trimethoxysilane 97%

Supplier: Ambeed
Warning

Synonyms: 3-Chloropropyltrimethoxysilane, 3-(Trimethoxysilyl)propylchloride, Trimethoxy(3-chloropropyl)silane

A164024-25G A164024-100G
77273-462EA 8 USD
77273-462 77550-042
(3-Chloropropyl)trimethoxysilane 97%
(3-Chloropropyl)trimethoxysilane

Precaución: for Research Use Only

Formula: ClH₂CCH₂CH₂Si(OCH₃)₃
MDL Number: MFCD00000997
CAS Number: 2530-87-2
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR