2-Chloro-6-methyl-benzoic acid methyl ester 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-chloro-6-methylbenzoate

A164017-100MG A164017-1G A164017-250MG A164017-5G
77259-718EA 12 USD
77259-718 77259-720 77259-722 77259-724
2-Chloro-6-methyl-benzoic acid methyl ester 95%
2-Chloro-6-methyl-benzoic acid methyl ester

Precaución: for Research Use Only

Formula: C₉H₉ClO₂
MDL Number: MFCD06204147
CAS Number: 99585-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR