1-(2-Bromoethoxy)-2-ethoxybenzene 98%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Ethoxyphenoxy)ethyl bromide

A163751-100G A163751-10G A163751-25G A163751-5G
77235-988EA 138.02 USD
77235-988 77235-990 77235-992 77235-994
1-(2-Bromoethoxy)-2-ethoxybenzene 98%
1-(2-Bromoethoxy)-2-ethoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrO₂
MW: 245.12 g/mol
MDL Number: MFCD02030483
CAS Number: 3259-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR