GZD824 98%

Supplier: Ambeed
Warning

Synonyms: Olverembatinib, 4-Methyl-N-[4-[(4-methyl-1-piperazinyl)methyl]-3-(trifluoromethyl)phenyl]-3-[2-(1H-pyrazolo[3,4-b]pyridin-5-yl)ethynyl]benzamide, 3-((1H-Pyrazolo[3,4-b]pyridin-5-yl)ethynyl)-4-methyl-N-(4-((4-methylpiperazin-1-yl)methyl)-3-(trifluoromethyl)phenyl)benzamide

A163539-1MG A163539-5MG
77235-960EA 8.24 USD
77235-960 77235-962
GZD824 98%
GZD824

Precaución: for Research Use Only

Formula: C₂₉H₂₇F₃N₆O
MW: 532.571 g/mol
MDL Number: MFCD26142930
CAS Number: 1257628-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR