N-Methyl-2-(pyridin-2-yl)-N-(2-(pyridin-2-yl)ethyl)ethanamine 97%

Supplier: Ambeed
Warning

Synonyms: Betahistine EP Impurity C

A163510-100MG A163510-250MG
77235-956EA 157.59 USD
77235-956 77235-958
N-Methyl-2-(pyridin-2-yl)-N-(2-(pyridin-2-yl)ethyl)ethanamine 97%
N-Methyl-2-(pyridin-2-yl)-N-(2-(pyridin-2-yl)ethyl)ethanamine

Precaución: for Research Use Only

Formula: C₁₅H₁₉N₃
MW: 241.33 g/mol
MDL Number: MFCD28157290
CAS Number: 5452-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR