Methyl-3-(phenylamino)propanoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-(phenylamino)propanoate

A163432-1G A163432-250MG
77286-536EA 55 USD
77286-536 77286-538
Methyl-3-(phenylamino)propanoate 98%
Methyl-3-(phenylamino)propanoate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MDL Number: MFCD00089334
CAS Number: 21911-84-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR