4-Chloro-3-cyclopropylpyridine hydrochloride 97%

Supplier: Ambeed
Warning

A163359-100MG A163359-1G A163359-250MG
77235-942EA 123 USD
77235-942 77235-944 77235-946
4-Chloro-3-cyclopropylpyridine hydrochloride 97%
4-Chloro-3-cyclopropylpyridine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₉Cl₂N
MDL Number: MFCD30345530
CAS Number: 1998216-32-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR