EI1 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Cyano-N-((4,6-dimethyl-2-oxo-1,2-dihydropyridin-3-yl)methyl)-1-(pentan-3-yl)-1H-indole-4-carboxamide, 6-cyano-N-[(1,2-dihydro-4,6-dimethyl-2-oxo-3-pyridinyl)methyl]-1-(1-ethylpropyl)-1H-indole-4-carboxamide
A163218-10MG
A163218-1MG
A163218-50MG
A163218-5MG
77235-920EA
53.56
USD
77235-920
77235-922
77235-924
77235-926
EI1 98%
EI1
Precaución: for Research Use Only
Formula:
C₂₃H₂₆N₄O₂ MW: 390.5 g/mol |
MDL Number:
MFCD25976740 CAS Number: 1418308-27-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...