3,4-Dimethoxysalicylic acid 95%

Supplier: Ambeed
Warning

A163210-1G A163210-250MG
77259-710EA 350 USD
77259-710 77259-712
3,4-Dimethoxysalicylic acid 95%
3,4-Dimethoxysalicylic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₅
MDL Number: MFCD00016498
CAS Number: 5653-46-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR