Methyl-3,5-dibromobenzoate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 3,5-dibromobenzoate
A163152-100G
A163152-10G
A163152-1G
A163152-25G
A163152-500G
A163152-5G
77235-908EA
83
USD
77235-908
77235-910
77235-912
77235-914
77235-916
77235-918
Methyl-3,5-dibromobenzoate 98%
Methyl 3,5-dibromobenzoate
Precaución: for Research Use Only
Formula:
C₈H₆Br₂O₂ |
MDL Number:
MFCD00082648 CAS Number: 51329-15-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...