2,3-Dihydrobenzo[b][1,4]dioxine-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A163137-100G A163137-10G A163137-1G A163137-25G A163137-5G
77259-700EA 167 USD
77259-700 77259-702 77259-704 77259-706 77259-708
2,3-Dihydrobenzo[b][1,4]dioxine-5-carboxylic acid 97%
2,3-Dihydrobenzo[b][1,4]dioxine-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₈O₄
MDL Number: MFCD00239415
CAS Number: 4442-53-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR