ω-Heptalactam 98%

Supplier: Ambeed
Warning

Synonyms: 2-Azacyclooctanone, Azocan-2-one, ω-Heptanolactam, ω-Enantholactam

A163131-100G A163131-1G A163131-250MG
77259-694EA 502.64 USD
77259-694 77259-696 77259-698
ω-Heptalactam 98%
omega-Heptalactam

Precaución: for Research Use Only

Formula: C₇H₁₃NO
MW: 127.19 g/mol
MDL Number: MFCD00003271
CAS Number: 673-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR