2,4,6-Tribromoresorcinol 98%

Supplier: Ambeed
Warning

A163071-1G A163071-250MG A163071-5G
77247-710EA 22 USD
77247-710 77247-712 77247-714
2,4,6-Tribromoresorcinol 98%
2,4,6-Tribromoresorcinol

Precaución: for Research Use Only

Formula: C₆H₃Br₃O₂
MDL Number: MFCD00009717
CAS Number: 2437-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR