5,6-Diamino-3-methyl-2-(methylthio)pyrimidin-4(3H)-one 97%

Supplier: Ambeed
Warning

A163025-100MG A163025-250MG
77247-700EA 7 USD
77247-700 77247-702
5,6-Diamino-3-methyl-2-(methylthio)pyrimidin-4(3H)-one 97%
5,6-Diamino-3-methyl-2-(methylthio)pyrimidin-4(3H)-one

Precaución: for Research Use Only

Formula: C₆H₁₀N₄OS
MW: 186.24 g/mol
MDL Number: MFCD02091208
CAS Number: 39008-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR