2,2,3,3,4,4,5,5-Octafluoro-1-pentanol 95%

Supplier: Ambeed
Warning

Synonyms: 1H,1H,5H-Octafluoro-1-pentanol

A163007-1000G A163007-100G A163007-25G A163007-500G A163007-5G
77247-690EA 66.95 USD
77247-690 77247-692 77247-694 77247-696 77247-698
2,2,3,3,4,4,5,5-Octafluoro-1-pentanol 95%
2,2,3,3,4,4,5,5-Octafluoro-1-pentanol

Precaución: for Research Use Only

Formula: C₅H₄F₈O
MW: 232.07 g/mol
MDL Number: MFCD00039631
CAS Number: 355-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR