3-(Boc-amino)-3-(4-fluorophenyl)-1-propanol 95%

Supplier: Ambeed
Warning

A162996-1G A162996-250MG A162996-5G
77286-516EA 68 USD
77286-516 77286-518 77286-520
3-(Boc-amino)-3-(4-fluorophenyl)-1-propanol 95%
3-(Boc-amino)-3-(4-fluorophenyl)-1-propanol

Precaución: for Research Use Only

Formula: C14H20FNO3
MDL Number: MFCD04115551
CAS Number: 862466-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR