Indole-3-carbaldehyd 98%

Supplier: Ambeed
Warning

A162961-100G A162961-10G A162961-500G A162961-25G A162961-1000G
77259-688EA 16 USD
77259-688 77259-690 77259-692 77320-008 77615-336
Indole-3-carbaldehyd 98%
Indole-3-carbaldehyd

Precaución: for Research Use Only

Formula: C₉H₇NO
MDL Number: MFCD00005622
CAS Number: 487-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR