Ethyl 6-(tert-butylamino)pyrimidine-4-carboxylate 97%

Supplier: Ambeed
Warning

A162944-100MG A162944-1G A162944-250MG A162944-5G
77273-416EA 11.33 USD
77273-416 77273-418 77273-420 77273-422
Ethyl 6-(tert-butylamino)pyrimidine-4-carboxylate 97%
Ethyl 6-(tert-butylamino)pyrimidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₇N₃O₂
MW: 223.27 g/mol
MDL Number: MFCD31619105
CAS Number: 2153472-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR