6-Methylpyrimidine-4(3H)-thione 97%

Supplier: Ambeed
Warning

A162848-100MG A162848-1G
77259-684EA 23 USD
77259-684 77259-686
6-Methylpyrimidine-4(3H)-thione 97%
6-Methylpyrimidine-4(3H)-thione

Precaución: for Research Use Only

Formula: C₅H₆N₂S
MW: 126.18 g/mol
MDL Number: MFCD08568504
CAS Number: 51793-97-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR