3-(1-Bromoethyl)benzotrifluoride 98%

Supplier: Ambeed
Danger

Synonyms: alpha-Methyl-3-(trifluoromethyl)benzyl bromide, 1-(1-Bromoethyl)-3-(trifluoromethyl)benzene

A162824-5G A162824-1G
77273-404EA 98.88 USD
77273-404 77748-094
3-(1-Bromoethyl)benzotrifluoride 98%
3-(1-Bromoethyl)benzotrifluoride

Precaución: for Research Use Only

Formula: C₉H₈BrF₃
MW: 253.0600 g/mol
MDL Number: MFCD00018050
CAS Number: 59770-96-6
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR