2-Chloro-3-cyanoquinoline 95%

Supplier: Ambeed
Warning

A162789-100MG A162789-1G A162789-250MG A162789-5G
77259-676EA 11 USD
77259-676 77259-678 77259-680 77259-682
2-Chloro-3-cyanoquinoline 95%
2-Chloro-3-cyanoquinoline

Precaución: for Research Use Only

Formula: C₁₀H₅ClN₂
MDL Number: MFCD08437566
CAS Number: 95104-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR