4-Bromo-2-(trifluoromethyl)phenol 98%

Supplier: Ambeed
Warning

A162773-1G A162773-25G A162773-5G
77286-508EA 12.36 USD
77286-508 77286-510 77286-512 77286-514
4-Bromo-2-(trifluoromethyl)phenol 98%
4-Bromo-2-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₄BrF₃O
MDL Number: MFCD05668981
CAS Number: 50824-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR