2-Amino-6-nitrobenzonitrile 95%

Supplier: Ambeed
Warning

A162741-100MG A162741-1G A162741-250MG
77259-670EA 72.1 USD
77259-670 77259-672 77259-674
2-Amino-6-nitrobenzonitrile 95%
2-Amino-6-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₅N₃O₂
MW: 163.1335 g/mol
MDL Number: MFCD08236739
CAS Number: 63365-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR