5-Hydroxysophthalic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Hydroxyisophthalic acid

A162694-100G A162694-10G A162694-25G A162694-500G A162694-1000G
77273-388EA 43 USD
77273-388 77273-390 77273-392 77273-394 77748-093
5-Hydroxysophthalic acid 98%
5-Hydroxysophthalic acid

Precaución: for Research Use Only

Formula: C₈H₆O₅
MDL Number: MFCD00002515
CAS Number: 618-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR