1-Chloro-10H-phenothiazine 97%

Supplier: Ambeed
Warning

A162500-1G A162500-250MG
77235-904EA 150 USD
77235-904 77235-906
1-Chloro-10H-phenothiazine 97%
1-Chloro-10H-phenothiazine

Precaución: for Research Use Only

Formula: C₁₂H₈ClNS
MDL Number: MFCD18447826
CAS Number: 1910-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR