LDC4297 98+%

Supplier: Ambeed
Warning

Synonyms: N-(2-(1H-Pyrazol-1-yl)benzyl)-8-isopropyl-2-(piperidin-3-yloxy)pyrazolo[1,5-a][1,3,5]triazin-4-amine

A162429-10MG A162429-1MG A162429-25MG A162429-50MG A162429-5MG
77273-368EA 57.68 USD
77273-368 77273-370 77273-372 77273-374 77273-376
LDC4297 98+%
LDC4297

Precaución: for Research Use Only

Formula: C₂₃H₂₈N₈O
MW: 432.52 g/mol
MDL Number: MFCD28411427
CAS Number: 1453834-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR