(R)-4-Benzyl-2-(quinolin-2-yl)-4,5-dihydrooxazole ≥98%, ee 99%

Supplier: Ambeed
Warning

A162312-100MG A162312-1G A162312-250MG
77259-654EA 61.8 USD
77259-654 77259-656 77259-658
(R)-4-Benzyl-2-(quinolin-2-yl)-4,5-dihydrooxazole ≥98%, ee 99%
(R)-4-Benzyl-2-(quinolin-2-yl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₉H₁₆N₂O
MW: 288.34 g/mol
CAS Number: 933992-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR