CeMMEC13 ≥98%

Supplier: Ambeed
Warning

Synonyms: N-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)-1-methyl-2-oxo-1,2-dihydroquinoline-4-carboxamide

A162180-10MG A162180-1MG A162180-25MG A162180-50MG A162180-5MG
77259-644EA 35.02 USD
77259-644 77259-646 77259-648 77259-650 77259-652
CeMMEC13 ≥98%
CeMMEC13

Precaución: for Research Use Only

Formula: C₁₉H₁₆N₂O₄
MW: 336.34 g/mol
MDL Number: MFCD30742989
CAS Number: 1790895-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR