2-(4-Benzyloxyphenyl)ethanol 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-(Benzyloxy)phenyl)ethanol

A161972-100G A161972-10G A161972-1G A161972-25G A161972-500G A161972-5G
77235-878EA 40 USD
77235-878 77235-880 77235-882 77235-884 77235-886 77615-332
2-(4-Benzyloxyphenyl)ethanol 98%
2-(4-Benzyloxyphenyl)ethanol

Precaución: for Research Use Only

Formula: C₁₅H₁₆O₂
MDL Number: MFCD00017532
CAS Number: 61439-59-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR