3-Chloro-5-hydroxybenzaldehyde 97%

Supplier: Ambeed
Warning

A161922-5G A161922-250MG A161922-1G
77273-356EA 184.37 USD
77273-356 77615-330 77748-090
3-Chloro-5-hydroxybenzaldehyde 97%
3-Chloro-5-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₅ClO₂
MW: 156.5664 g/mol
MDL Number: MFCD08234659
CAS Number: 1829-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR