2-Fluoro-3-iodopyridine 98%

Supplier: Ambeed
Warning

A161909-100G A161909-10G A161909-1G A161909-25G A161909-5G
77273-346EA 198.79 USD
77273-346 77273-348 77273-350 77273-352 77273-354
2-Fluoro-3-iodopyridine 98%
2-Fluoro-3-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₃FIN
MW: 222.99 g/mol
MDL Number: MFCD03095287
CAS Number: 113975-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR