Trimethyl ethane-1,1,2-tricarboxylate 95%

Supplier: Ambeed
Warning

A161878-100G A161878-10G A161878-1G A161878-25G
77259-632EA 35 USD
77259-632 77259-634 77259-636 77259-638 77259-640
Trimethyl ethane-1,1,2-tricarboxylate 95%
Trimethyl ethane-1,1,2-tricarboxylate

Precaución: for Research Use Only

Formula: C₈H₁₂O₆
MW: 204.1800 g/mol
MDL Number: MFCD00015619
CAS Number: 40967-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR