4-Benzyl-L-aspartate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
L-Aspartic acid beta-benzyl ester, β-Benzyl L-aspartate, H-Asp(obzl)-OH, beta-Benzyl L-aspartate, L-Aspartic acid 4-benzyl ester, L-Aspartic Acid 4-Benzyl Ester, beta-Benzyl L-Aspartate, H-Asp(OBzl)-OH, 4-Benzyl L-Aspartate, H-Asp(OBn)-OH, L-Aspartic Acid beta-Benzyl Ester, L-Aspartic acid β-benzyl ester
A161792-100G
A161792-10G
A161792-25G
A161792-5G
A161792-500G
77273-338EA
35.02
USD
77273-338
77273-340
77273-342
77273-344
77748-087
4-Benzyl-L-aspartate 97%
4-Benzyl-L-aspartate
Precaución: for Research Use Only
Formula:
C₁₁H₁₃NO₄ MW: 223.23 g/mol |
MDL Number:
MFCD00037208 CAS Number: 2177-63-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...