4-Hydroxybenzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: p-Hydroxybenzoic Acid

A161723-1000G A161723-100G A161723-10G A161723-25G A161723-500G
77286-468EA 25.75 USD
77286-468 77286-470 77286-472 77286-474 77286-476
4-Hydroxybenzoic acid 98%
4-Hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₆O₃
MW: 138.12 g/mol
MDL Number: MFCD00002547
CAS Number: 99-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR