4-Fluoro-3-(trifluoromethyl)phenylacetic acid 98%

Supplier: Ambeed
Warning

A161711-1G A161711-25G A161711-5G
77286-462EA 11.33 USD
77286-462 77286-464 77286-466
4-Fluoro-3-(trifluoromethyl)phenylacetic acid 98%
4-Fluoro-3-(trifluoromethyl)phenylacetic acid

Precaución: for Research Use Only

Formula: C₉H₆F₄O₂
MW: 222.14 g/mol
MDL Number: MFCD00061190
CAS Number: 220227-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR