2-Acetyl-4-chlorothiophene 97%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Chlorothiophen-2-yl)ethanone, 1-(4-Chloro-2-thienyl)-1-ethanone

A161542-10G A161542-1G A161542-250MG A161542-5G
77247-660EA 134.93 USD
77247-660 77247-662 77247-664 77247-666
2-Acetyl-4-chlorothiophene 97%
2-Acetyl-4-chlorothiophene

Precaución: for Research Use Only

Formula: C₆H₅ClOS
MW: 160.62 g/mol
MDL Number: MFCD00082791
CAS Number: 34730-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR