Rhodanine-N-acetic acid 97%

Supplier: Ambeed
Warning

A161514-100G A161514-10G A161514-1G A161514-25G A161514-5G A161514-500G
77286-444EA 53 USD
77286-444 77286-446 77286-448 77286-450 77286-452 77567-354
Rhodanine-N-acetic acid 97%
Rhodanine-N-acetic acid

Precaución: For research use only.

Formula: C₅H₅NO₃S₂
MDL Number: MFCD00005491
CAS Number: 5718-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR