2-Nitro-4-(trifluoromethyl)benzenesulfonyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 2-Nitro-4-(trifluoromethyl)benzene-1-sulphonyl chloride

A161409-250MG A161409-5G
77273-322EA 15.45 USD
77273-322 77273-324
2-Nitro-4-(trifluoromethyl)benzenesulfonyl chloride 98%
2-Nitro-4-(trifluoromethyl)benzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₇H₃ClF₃NO₄S
MW: 289.62 g/mol
MDL Number: MFCD00013456
CAS Number: 837-95-6
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR