3-Bromochromone 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-4H-chromen-4-one

A161403-250MG A161403-5G
77273-318EA 16.48 USD
77273-318 77273-320
3-Bromochromone 98%
3-Bromochromone

Precaución: for Research Use Only

Formula: C₉H₅BrO₂
MW: 225.05 g/mol
MDL Number: MFCD00017337
CAS Number: 49619-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR