tert-Butyl 2-(6-chloropyridin-3-yl)pyrrolidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A161315-100MG A161315-250MG
77235-872EA 88.58 USD
77235-872 77235-874
tert-Butyl 2-(6-chloropyridin-3-yl)pyrrolidine-1-carboxylate 97%
tert-Butyl 2-(6-chloropyridin-3-yl)pyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₉ClN₂O₂
MW: 282.76 g/mol
MDL Number: MFCD20530216
CAS Number: 1352534-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR